ContactUsLinkComponent
EnzyChrom™ Triglyceride Assay Kit, BioAssay Systems
EnzyChrom™ Triglyceride Assay Kit, BioAssay Systems
# de catalogue 75878-176
Fournisseur:  BIOASSAY SYSTEMS
Numéro CAS:  
EnzyChrom* Triglyceride Assay Kit 200 tests
EnzyChrom™ Triglyceride Assay Kit, BioAssay Systems
# de catalogue 75878-176
Fournisseur:  BIOASSAY SYSTEMS
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Triglyceride Assay Kit
  • Size:
    200 assays
  • Method:
    OD570 nm
  • Sample:
    Serum, plasma, etc.
  • Processing time:
    30 minutes
  • Cat. no.:
    75878-176
  • Supplier No.:
    ETGA-200
Spécifications
A propos de cet article
For quantitative determination of triglyceride and evaluation of drug effects on triglyceride metabolism.
  • Linear detection range 0.01 - 1.0 mmol/L (0.88 - 88.5 mg/dL) triglyceride
  • Convenient room temperature high-throughput assay
Triglyceride, also known as Triacyltriglyceride or Triacyl-glyceride, is the main constituent in vegetable oil and animal fats
BioAssay Systems’ triglyceride assay uses a single Working Reagent that combines triglyceride hydrolysis and glycerol determination in one step, in which a dye reagent is oxidized to form a colored product. The color intensity at 570nm is directly proportional to triglyceride concentration in the sample.
Triglycerides play an important role as energy sources and transporters of dietary fat. In the human body, high levels of triglycerides in the bloodstream have been linked to atherosclerosis, heart disease and pancreatitis. Simple, direct and automation-ready. Procedures for measuring triglyceride concentrations find wide applications in research and drug discovery.
Customers Who Bought This Also Bought