ContactUsLinkComponent
EnzyChrom™ Superoxide Dismutase Assay Kit, BioAssay Systems
EnzyChrom™ Superoxide Dismutase Assay Kit, BioAssay Systems
# de catalogue 75878-172
Fournisseur:  BIOASSAY SYSTEMS
Numéro CAS:  
EnzyChrom* Superoxide Dismutase Assay Kit 100 tests
EnzyChrom™ Superoxide Dismutase Assay Kit, BioAssay Systems
# de catalogue 75878-172
Fournisseur:  BIOASSAY SYSTEMS
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Superoxide Dismutase Assay Kit
  • Size:
    100 assays
  • Method:
    OD440 nm
  • Sample:
    Blood, cell, tissue etc.
  • Processing time:
    One hour
  • Cat. no.:
    75878-172
  • Supplier No.:
    ESOD-100
Spécifications
A propos de cet article
For quantitative determination of superoxide dismutase (SOD) enzyme activity.
  • Linear detection range of 0.05 - 3 U/mL SOD
  • No wash and reagent transfer steps are involved
  • Homogeneous "mix-incubate-measure" type high-throughput assay
Superoxide dismutases (SOD, EC1.15.1.1) are enzymes that catalyze the dismutation of superoxide into O2 and H2O2
BioAssay Systems’ SOD assay provides a convenient colorimetric means for the quantitative determination of SOD enzyme activity in biological samples. In the assay, superoxide (O2-) is provided by xanthine oxidase (XO) catalyzed reaction. O2- reacts with a WST-1 dye to form a colored product. SOD scavenges the O2- thus less O2- is available for the chromogenic reaction. The color intensity (OD440nm) is used to determine the SOD activity in a sample.
They are an important antioxidant defense in all cells exposed to O2. There are three major families of superoxide dismutase: Cu/Zn, Fe/Mn, and the Ni type. Aberrant SOD activities have been linked to diseases such as amyotrophic lateral sclerosis, perinatal lethality, neural disorders and cancer.
Customers Who Bought This Also Bought