ContactUsLinkComponent
EnzyChrom™ Sialic acid Assay Kit, BioAssay Systems
EnzyChrom™ Sialic acid Assay Kit, BioAssay Systems
# de catalogue 75878-170
Fournisseur:  BIOASSAY SYSTEMS
Numéro CAS:  
EnzyChrom* Sialic Acid Assay Kit 100 tests
EnzyChrom™ Sialic acid Assay Kit, BioAssay Systems
# de catalogue 75878-170
Fournisseur:  BIOASSAY SYSTEMS
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Sialic acid Assay Kit
  • Size:
    100 assays
  • Method:
    OD570 nm, or FL530/585 nm
  • Sample:
    Biological
  • Processing time:
    Two hours
  • Cat. no.:
    75878-170
  • Supplier No.:
    ESLA-100
Spécifications
A propos de cet article
For quantitative determination of free and total sialic acid (NANA) concentration.
  • Linear detection range 20 - 1000 μM sialic acid for colorimetric assays and 2 - 100 μM for fluorimetric assays
  • Can detect both free and total sialic acid
Sialic acid is a general name for nine carbon acidic sugars with N- or O-substituted derivatives
BioAssay Systems’ sialic acid assay uses a single Working Reagent that combines NANA aldolase, pyruvate oxidase and hydrogen peroxide determination in one step. The color intensity of the reaction product at 570nm or fluorescence intensity at λex/em = 530/585nm is directly proportional to sialic acid concentration in the sample.
The most common member of these sugars is N-acetylneuraminic acid (NANA). Sialic acid is widely distributed throughout mammalian tissues and fluids including serum. Sialylated oligosaccharides have been shown to exhibit antiviral properties and are also known to influence blood coagulation and cholesterol levels. The Sialic acid level in body fluids is also an important marker for diagnosing cancer. Simple, direct and automation-ready. Procedures for measuring sialic acid concentrations find wide applications in research and drug discovery.
Customers Who Bought This Also Bought