ContactUsLinkComponent
EnzyChrom™ Sorbitol Assay Kit, BioAssay Systems
EnzyChrom™ Sorbitol Assay Kit, BioAssay Systems
# de catalogue 75878-168
Fournisseur:  BIOASSAY SYSTEMS
Numéro CAS:  
EnzyChrom* Sorbitol Assay Kit 100 tests
EnzyChrom™ Sorbitol Assay Kit, BioAssay Systems
# de catalogue 75878-168
Fournisseur:  BIOASSAY SYSTEMS
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Sorbitol Assay Kit
  • Size:
    100 assays
  • Method:
    OD565 nm
  • Sample:
    Biological (e.g. blood), food, beverage and agriculture samples.
  • Processing time:
    30 minutes
  • Cat. no.:
    75878-168
  • Supplier No.:
    ESBT-100
Spécifications
A propos de cet article
For quantitative determination of D-sorbitol concentration.
  • Linear detection range 5 - 1000 μM D-sorbitol.
  • Convenient room temperature assay
  • No 37°C incubator required
  • Can be readily automated to test thousands of samples per day
The mitogen-activated protein kinase (MAPK/ERK) pathway plays a key role in cell proliferation, differentiation and migration
BioAssay Systems’ cell-based ELISA measures dually phosphorylated ERK1/2 in whole cells. This simple and efficient assay eliminates the need for cell lysate preparation and normalizes the signal to the total protein content. The assay is a useful tool to study the MAP kinase signaling pathway. In this assay, cells are grown in 96-well plates and treated with ligands or drugs. Cells are then fixed and permeabilized in the wells. ERK1/2 phosphorylation (pERK) is measured using a HRP-conjugated antibody specific for phosphorylated ERK1/2.
Stimulation by mitogens eventually leads to phosphorylation of ERK1 (T202/Y204) and ERK2 (T185/Y187). The MAPK/ERK cascade presents many interesting drug targets for the development of cancer therapies.
Customers Who Bought This Also Bought