BioMasher® II Micro Tissue Homogenizers, Closed System, Disposable, Kimble Chase
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
BioMasher® II is a micro sized closed system tissue homogenizers having a pestle with a molded-in shaft guard to minimize potential exposure to hazardous materials.
- Clear polypropylene tube
- Polyacetal pestle
The abrasive surfaces on the pestle tip and inner tube area ensure efficient grinding of samples. Ideal for homogenizing of animal tissues and organs as well as plant materials, insects, DNA, RNA, proteins, yeasts and enzymes.
Customers Who Bought This Also Bought