ContactUsLinkComponent
EnzyChrom™ Phospholipid Assay Kit, BioAssay Systems
EnzyChrom™ Phospholipid Assay Kit, BioAssay Systems
# de catalogue 75878-160
Fournisseur:  BIOASSAY SYSTEMS
Numéro CAS:  
EnzyChrom* Phospholipid Assay Kit 100tests
EnzyChrom™ Phospholipid Assay Kit, BioAssay Systems
# de catalogue 75878-160
Fournisseur:  BIOASSAY SYSTEMS
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Phospholipid Assay Kit
  • Size:
    100 assays
  • Method:
    OD570 nm, or FL530/585 nm
  • Sample:
    Biological
  • Processing time:
    20 minutes
  • Cat. no.:
    75878-160
  • Supplier No.:
    EPLP-100
Spécifications
A propos de cet article
For quantitative determination of phospholipid and evaluation of drug effects on its metabolism.
  • Linear detection range colorimetric assay 3 - 200 μM, fluorimetric assay 0.6 - 20 μM phospholipid
Phospholipids are a class of lipids which constitute a major component of cell membranes and play important roles in signal transduction
BioAssay Systems’ method provides a simple, direct and high-throughput assay for measuring choline-containing phospholipids in biological samples. In this assay, phospholipids (such as lecithin, lysolecithin and sphingomyelin) are enzymatically hydrolyzed to choline which is determined using choline oxidase and a H2O2 specific dye. The optical density of the pink colored product at 570nm or fluorescence intensity (530/585 nm) is directly proportional to the phospholipid concentration in the sample.
Most phospholipids contain one diglyceride, a phosphate group, and one choline.
Customers Who Bought This Also Bought