ContactUsLinkComponent
EnzyChrom™ Oxalate Assay Kit, BioAssay Systems
EnzyChrom™ Oxalate Assay Kit, BioAssay Systems
# de catalogue 75878-206
Fournisseur:  BIOASSAY SYSTEMS
Numéro CAS:  
EnzyChrom* Oxalate Assay Kit 100 tests
EnzyChrom™ Oxalate Assay Kit, BioAssay Systems
# de catalogue 75878-206
Fournisseur:  BIOASSAY SYSTEMS
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Oxalate Assay Kit
  • Size:
    100 assays
  • Method:
    OD595 nm
  • Sample:
    Urine, animal and plant tissue samples
  • Processing time:
    10 minutes
  • Cat. no.:
    75878-206
  • Supplier No.:
    EOXA-100
Spécifications
A propos de cet article
For quantitative determination of oxalate (oxalic acid) in urine and other samples.
  • Linear detection range 20 - 1500 µM oxalate
  • Sample pre-treatment is faster and easier than using activated carbon in competitor’s assay kits
  • Can be automated to test thousands of samples per day
Oxalate or Oxalic Acid is a metabolic breakdown product of the Kreb’s Cycle in eukaryotes, and the glyoxylate cycle in other microorganisms
BioAssay Systems’ oxalate assay kit uses a single Working Reagent that combines the oxalate oxidase reaction and color reaction in one step. The change in color intensity of the reaction product at 595 nm is directly proportional to oxalate in the sample.
It can be found in the urine of humans and other mammals. Oxalate concentration can be used as a measure of kidney function where a high level of oxalate is an indicator for kidney stones which are primarily made of the insoluble salt calcium oxalate. Measuring oxalate is more accurate than measuring calcium as a marker for kidney stones because calcium is excreted at high concentrations even in normal urine. Simple and high-throughput assays for measuring oxalate concentration find wide applications.
Customers Who Bought This Also Bought