Spécifications
- Description:Ammonia/Ammonium Assay Kit
- Size:100 assays
- Method:OD340 nm, or FL360/450 nm
- Sample:Serum, plasma, urine, saliva, cell culture etc.
- Processing time:30 minutes
- Cat. no.:75878-156
- Supplier No.:ENH3-100
Spécifications
A propos de cet article
For quantitative colorimetric/fluorometric determination of ammonia/ammonium concentrations.
- Linear detection range 24 to 1000 μM ammonia
- Simple "mix-and-measure" type assay
Ammonia (NH3) or its ion form ammonium (NH4+) is an important source of nitrogen for living systems
BioAssay Systems’ ammonia assay is designed to directly measure NH3 and NH4+. In this assay, NADH is converted to NAD+ in the presence of NH3, ketoglutarate and glutamate dehydrogenase. The decrease in optical density at 340 nm or fluorescence intensity at λex/em = 360/450 nm is directly proportionate to the NH3 concentration in the sample.
It is synthesized through amino acid metabolism and is toxic when present at high concentrations. In the liver, ammonia is converted to urea through the urea cycle. Elevated levels of ammonia in the blood (hyperammonemia) have been found in liver dysfunction (cirrhosis), while hypoammonemia has been associated with defects in the urea cycle enzymes (e.g. ornithine transcarbamylase). Simple, direct and automation-ready. Procedures for measuring NH3 are popular in research and drug discovery.