ContactUsLinkComponent
EnzyChrom™ Ammonia/Ammonium Assay Kit, BioAssay Systems
EnzyChrom™ Ammonia/Ammonium Assay Kit, BioAssay Systems
# de catalogue 75878-156
Fournisseur:  BIOASSAY SYSTEMS
Numéro CAS:  
EnzyChrom* Ammonia/Ammonium Assay Kit 100 tests
EnzyChrom™ Ammonia/Ammonium Assay Kit, BioAssay Systems
# de catalogue 75878-156
Fournisseur:  BIOASSAY SYSTEMS
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Ammonia/Ammonium Assay Kit
  • Size:
    100 assays
  • Method:
    OD340 nm, or FL360/450 nm
  • Sample:
    Serum, plasma, urine, saliva, cell culture etc.
  • Processing time:
    30 minutes
  • Cat. no.:
    75878-156
  • Supplier No.:
    ENH3-100
Spécifications
A propos de cet article
For quantitative colorimetric/fluorometric determination of ammonia/ammonium concentrations.
  • Linear detection range 24 to 1000 μM ammonia
  • Simple "mix-and-measure" type assay
Ammonia (NH3) or its ion form ammonium (NH4+) is an important source of nitrogen for living systems
BioAssay Systems’ ammonia assay is designed to directly measure NH3 and NH4+. In this assay, NADH is converted to NAD+ in the presence of NH3, ketoglutarate and glutamate dehydrogenase. The decrease in optical density at 340 nm or fluorescence intensity at λex/em = 360/450 nm is directly proportionate to the NH3 concentration in the sample.
It is synthesized through amino acid metabolism and is toxic when present at high concentrations. In the liver, ammonia is converted to urea through the urea cycle. Elevated levels of ammonia in the blood (hyperammonemia) have been found in liver dysfunction (cirrhosis), while hypoammonemia has been associated with defects in the urea cycle enzymes (e.g. ornithine transcarbamylase). Simple, direct and automation-ready. Procedures for measuring NH3 are popular in research and drug discovery.
Customers Who Bought This Also Bought