Rejuvenator® Polyurethane Squared Surface Runners, Wearwell®
Fournisseur: Wearwell
Certificats
A propos de cet article
Rejuvenator® runners deliver unmatched anti-fatigue benefits and long-term performance.
- 100% urethane with thick outer wear layer
- Deep positive lock keeps seams secure
- Quickly form endless lengths
- Molded-in safety bevels
- Meets USP 797 compliance standards
- Ideally suited for dry environments
Rejuvenator® Runners provide the option to create 2' or 3' wide runs of ergonomic matting. The mats are available in Ends and Centers, allowing users to create the specific length of run. The molded-in ramps reduce installation time and costs because there is no edging to attach.
Certifications: ASTM F1677. Dry: COF = 0.75. Wet: COF = 0.28.
Customers Who Bought This Also Bought