ContactUsLinkComponent
EnzyChrom™ Ketone body Assay Kit, BioAssay Systems
EnzyChrom™ Ketone body Assay Kit, BioAssay Systems
# de catalogue 75878-144
Fournisseur:  BIOASSAY SYSTEMS
Numéro CAS:  
EnzyChrom* Ketone Body Assay Kit 100 tests
EnzyChrom™ Ketone body Assay Kit, BioAssay Systems
# de catalogue 75878-144
Fournisseur:  BIOASSAY SYSTEMS
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Ketone Body Assay Kit
  • Size:
    100 assays
  • Method:
    OD340 nm
  • Sample:
    Serum, plasma, urine etc.
  • Processing time:
    30 minutes
  • Cat. no.:
    75878-144
  • Supplier No.:
    EKBD-100
Spécifications
A propos de cet article
For quantitative determination of ketone body (acetoacetate and 3-hydroxybutyrate) and evaluation of drug effects on its metabolism.
  • Linear detection range of 0.12 to 8 mM for each ketone body in 96-well plate assay
  • The procedure involves adding a single working reagent, and reading the optical density at room temperature
  • Can be automated to test many samples per day
Ketone bodies (acetoacetic acid and 3-hydroxybutyric acid) are produced in the liver mainly from oxidation of fatty acids, and are normally present at low concentrations in urine and blood
BioAssay Systems’ EnzyChrom™ ketone body assay is based on 3-hydroxybutyrate dehydrogenase catalyzed reactions, in which the change in NADH absorbance, measured at 340nm, is directly related to the AcAc and BOH concentrations,
Increased ketone concentrations in the blood may lead to metabolic acidosis, which has been associated with diabetes, childhood hypo-glycaemia, growth hormone deficiency, alcohol or salicylate intoxication and inborn errors of metabolism. Simple, direct and automation-ready. Procedures for measuring acetoacetic acid (AcAc) and 3-hydroxybutyric acid (BOH) are very desirable.
Customers Who Bought This Also Bought