ContactUsLinkComponent
EnzyChrom™ Invertase/Sucrase Assay Kit, BioAssay Systems
EnzyChrom™ Invertase/Sucrase Assay Kit, BioAssay Systems
# de catalogue 75878-142
Fournisseur:  BIOASSAY SYSTEMS
Numéro CAS:  
EnzyChrom* Invertase Assay Kit 100 tests
EnzyChrom™ Invertase/Sucrase Assay Kit, BioAssay Systems
# de catalogue 75878-142
Fournisseur:  BIOASSAY SYSTEMS
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Invertase/Sucrase Assay Kit
  • Size:
    100 assays
  • Method:
    OD570 nm, or FL530/585 nm
  • Sample:
    Biological, environment (soil) etc.
  • Processing time:
    40 minutes
  • Cat. no.:
    75878-142
  • Supplier No.:
    EIVT-100
Spécifications
A propos de cet article
For quantitative determination of invertase/sucrase activity.
  • Safe
  • Non-radioactive assay
  • "Mix-incubate-measure" type assay. No wash and reagent transfer steps are involved
  • Can be readily automated to test thousands of samples per day
Invertase (b-fructofuranosidase, EC 3.2.1.26) is an enzyme that catalyzes the hydrolysis of sucrose to fructose and glucose
BioAssay Systems’ Invertase Assay Kit provides a convenient and ultra-sensitive colorimetric and fluorimetric means to measure invertase activity. In the assay, invertase cleaves sucrose, resulting in the formation of fructose and glucose, which is determined by a colorimetric (570nm) or fluorimetric method (λex/em = 530/585nm). The assay is simple, sensitive, stable and high-throughput adaptable.
Invertases cleave at the O-C(fructose) bond, whereas a related enzyme sucrase (EC 3.2.1.48) cleaves at the O-C(glucose) bond. A wide range of microorganisms produce invertase and can, thus, utilize sucrose as a nutrient. Invertase assay finds wide applications in environmental (e.g. soil), agricultural and food (confectionery) industry.
Customers Who Bought This Also Bought