To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Description:Glucose Oxidase Assay Kit
- Size:100 assays
- Method:OD570 nm, FL530/585 nm
- Sample:Cell/tissue lysate, cell culture media etc.
- Processing time:20 minutes
- Cat. no.:75878-134
- Supplier No.:EGOX-100
Spécifications
A propos de cet article
For quantitative determination of glucose oxidase activity and evaluation of drug effects on its metabolism.
- Linear detection range 0.02 to 10 U/L glucose oxidase for colorimetric assays and 0.002 to 1.5 U/L for fluorimetric assays
- Homogeneous high-throughput assay
Glucose oxidase catalyzes the oxidation of glucose from D-glucose to D-glucono-δ-lactone
BioAssay Systems’ glucose oxidase assay kit uses a single Working Reagent that combines the glucose oxidase reaction and color reaction in one step. The change in color intensity of the reaction product at 570 nm or fluorescence intensity at λex/em = 530/585 nm is directly proportional to glucose oxidase activity in the sample.
Physiologically, it aids in the breakdown of glucose into smaller metabolites. It is widely used in electrochemical glucose sensors designed for diabetes patients. Simple, direct and high-throughput assays for measuring glucose oxidase activity find wide applications in research and drug discovery.