ContactUsLinkComponent
EnzyChrom™ Glucose-6-Phosphate Assay Kit, BioAssay Systems
EnzyChrom™ Glucose-6-Phosphate Assay Kit, BioAssay Systems
# de catalogue 75878-112
Fournisseur:  BIOASSAY SYSTEMS
Numéro CAS:  
EnzyChrom* Glucose-6-Phosphate Assay Kit 100 tests
EnzyChrom™ Glucose-6-Phosphate Assay Kit, BioAssay Systems
# de catalogue 75878-112
Fournisseur:  BIOASSAY SYSTEMS
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Glucose-6-Phosphate Assay Kit
  • Size:
    100 assays
  • Method:
    OD460 nm
  • Sample:
    Plasma, serum, tissue and culture media etc.
  • Processing time:
    20 minutes
  • Cat. no.:
    75878-112
  • Supplier No.:
    EG6P-100
Spécifications
A propos de cet article
For quantitative determination of glucose-6-phosphate in biological samples.
  • Linear detection range 10 to 1000 μM G6P
  • Homogeneous "mix-incubate-measure" type high-throughput assay
Glucose-6-Phosphate (g6p) is glucose sugar phosphorylated on carbon 6
BioAssay Systems’ Glucose-6-Phosphate Assay Kit provides a simple, and automation-ready procedure for measuring G6P concentration. G6P is oxidized by glucose-6-phosphate dehydrogenase and the formed NADPH is coupled to the formazan (WST-8) chromogen. The intensity of the product color, measured at 460 nm, is proportional to the G6P concentration in the sample.
Most of the glucose entering cells is phosphorylated to G6P. G6P has three primary fates within the cell. It lies at the start of two major metabolic pathways: glycolysis and the pentose phosphate pathway. In addition to these metabolic pathways, glucose 6-phosphate may also be converted to glycogen or starch for storage.
Customers Who Bought This Also Bought