To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Description:Acetate Assay Kit
- Size:100 assays
- Method:OD570 nm, or FL530/585 nm
- Sample:Serum, plasma, food, agriculture and environment
- Processing time:30 minutes
- Brand name:EnzyChrom™
- Cat. no.:75878-346
- Supplier No.:EOAC-100
Spécifications
A propos de cet article
For quantitative determination of acetic acid or acetate and evaluation of drug effects on acetate metabolism
- Use as little as 10 μL samples. Detection range: 0.20 to 20 mM acetate for colorimetric assays and 0.13 to 2 mM for fluorimetric assays.
- Method: OD570nm, or FL530/585nm
- Samples: Serum, plasma, food, agriculture and environment
Acetate is a common anion and fundamental to all forms of life. When bound to coenzyme A, it is central to the metabolism of carbohydrates and fats. It is acid form, acetic acid, is produced and excreted by acetic acid bacteria, such as Acetobacter genus and Clostridium acetobutylicum, which are found universally in foodstuffs, water, and soil. Acetic acid is also a component of the vaginal lubrication of humans and other primates, where it appears to serve as a mild antibacterial agent. Acetic acid is the main component of vinegar, and extensively used in food, dyes, paints, glue and synthetic fibres. BioAssay Systems’ assay uses enzyme-coupled reactions to form a colored, fluorescent product. The color absorbance at 570nm or fluorescence intensity at 530nm/585nm is directly proportional to the acetate concentration in the sample.