ContactUsLinkComponent
EnzyFluo™ Lactate Assay Kit, BioAssay Systems
EnzyFluo™ Lactate Assay Kit, BioAssay Systems
# de catalogue 75878-088
Fournisseur:  BIOASSAY SYSTEMS
Numéro CAS:  
EnzyFluo* L-Lactate Assay Kit 100 tests
EnzyFluo™ Lactate Assay Kit, BioAssay Systems
# de catalogue 75878-088
Fournisseur:  BIOASSAY SYSTEMS
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Lactate Assay Kit
  • Size:
    100 assays
  • Method:
    FL530/585 nm
  • Sample:
    Serum, plasma, cell culture media etc.
  • Processing time:
    60 minutes
  • Cat. no.:
    75878-088
  • Supplier No.:
    EFLLC-100
Spécifications
A propos de cet article
For quantitative fluorimetric determination of L-lactate (L-lactic acid) and evaluation of drug effects on its metabolism.
  • Linear detection range 1 - 50 μM L-lactate in 96-well plate assay
  • Can be readily automated to test thousands of samples per day
Lactate is generated by lactate dehydrogenase (LDH) under hypoxic or anaerobic conditions
BioAssay Systems’ EnzyFluo™ lactate assay kit is based on lactate dehydrogenase catalyzed oxidation of lactate, in which the formed NADH reduces a probe into a highly fluorescent product. The fluorescence intensity of this product, measured at λex/em = 530/585 nm, is proportional to the lactate concentration in the sample.
Monitoring lactate levels is, therefore, a good indicator of the balance between tissue oxygen demand and utilization and is useful when studying cellular and animal physiology. Simple, direct and automation-ready. Procedures for measuring lactate concentration are very desirable.
Customers Who Bought This Also Bought