ContactUsLinkComponent
EnzyFluo™ Isocitrate Assay Kit, BioAssay Systems
EnzyFluo™ Isocitrate Assay Kit, BioAssay Systems
# de catalogue 75878-086
Fournisseur:  BIOASSAY SYSTEMS
Numéro CAS:  
EnzyFluo* Isocitrate Assay Kit 100tests
EnzyFluo™ Isocitrate Assay Kit, BioAssay Systems
# de catalogue 75878-086
Fournisseur:  BIOASSAY SYSTEMS
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Isocitrate Assay Kit
  • Size:
    100 assays
  • Method:
    FL530/585 nm
  • Sample:
    Food, beverage, biological Samples (e.g. cell lysate, tissue homogenate, serum, etc.).
  • Processing time:
    10 minutes
  • Cat. no.:
    75878-086
  • Supplier No.:
    EFIC-100
Spécifications
A propos de cet article
For quantitative fluorimetric determination of isocitrate and evaluation of drug effects on its metabolism.
  • Linear detection range 0.6 to 500 μM for 10 min reaction
  • Homogeneous "mix-incubate-measure" type high-throughput assay
Isocitrate (Isocitric acid) is a substrate in the citric acid (TCA) cycle
BioAssay Systems’ isocitrate assay measures the NADPH generated from the oxidation of isocitrate. The NADPH reduces a probe into a highly fluorescent product. The fluorescence intensity of this product, measured at λex/em = 530/585 nm, is proportional to the isocitrate concentration in the sample.
Isocitrate is formed by the isomerization of citrate catalyzed by the enzyme aconitase. Isocitrate is oxidized by isocitrate dehydrogenase producing α-ketoglutarate and generating NADPH. Isocitrate is commonly found in many fruits and vegetables and their processed products. Industrially, isocitrate is used as a marker to identify the quality and purity of fruit juices.
Customers Who Bought This Also Bought