ContactUsLinkComponent
EnzyChrom™ Fatty acid Assay Kit, BioAssay Systems
EnzyChrom™ Fatty acid Assay Kit, BioAssay Systems
# de catalogue 75878-084
Fournisseur:  BIOASSAY SYSTEMS
Numéro CAS:  
EnzyChrom* Free Fatty Acid Assay Kit 100tests
EnzyChrom™ Fatty acid Assay Kit, BioAssay Systems
# de catalogue 75878-084
Fournisseur:  BIOASSAY SYSTEMS
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Fatty acid Assay Kit
  • Size:
    100 assays
  • Method:
    OD570 nm or FL530/585 nm
  • Sample:
    Serum, plasma, urine, saliva, milk, cell cultures, food, agriculture etc.
  • Processing time:
    30 minutes
  • Cat. no.:
    75878-084
  • Supplier No.:
    EFFA-100
Spécifications
A propos de cet article
For quantitative determination of fatty acid and evaluation of drug effects on its metabolism.
  • Linear detection range colorimetric assay 7 - 1000 μM, fluorimetric assay 7 - 100 μM fatty acid
  • Convenient "mix-and-read" high-throughput assay
Fatty acids are aliphatic monocarboxylic acids that are ubiquitously found in animal or vegetable fat, oil and wax
BioAssay Systems’ method provides a simple, one-step and high-throughput assay for measuring free fatty acids. In this assay, free fatty acids are enzymatically converted to acyl-CoA and subsequently to H2O2. The resulting H2O2 reacts with a specific dye to form a pink colored product. The optical density at 570nm or fluorescence intensity (530/585 nm) is directly proportional to the free fatty acid concentration in the sample.
Fatty Acids play important roles in cellular synthesis, energy metabolism and are implicated in diverse disorders such as diabetes mellitus, sudden infant death syndrome and Reye Syndrome.
Customers Who Bought This Also Bought