ContactUsLinkComponent
EnzyChrom™ Creatine Assay Kit, BioAssay Systems
EnzyChrom™ Creatine Assay Kit, BioAssay Systems
# de catalogue 75878-078
Fournisseur:  BIOASSAY SYSTEMS
Numéro CAS:  
EnzyChrom* Creatine Assay Kit 100tests
EnzyChrom™ Creatine Assay Kit, BioAssay Systems
# de catalogue 75878-078
Fournisseur:  BIOASSAY SYSTEMS
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Creatine Assay Kit
  • Size:
    100 assays
  • Method:
    OD570 nm, or FL530/585 nm
  • Sample:
    Serum, plasma, urine, saliva etc.
  • Processing time:
    30 minutes
  • Cat. no.:
    75878-078
  • Supplier No.:
    ECRT-100
Spécifications
A propos de cet article
For quantitative determination of creatine and evaluation of drug effects on creatine metabolism.
  • Linear detection range 4 to 1000 μM (colorimetric assay) or 0.5 to 50 μM (fluorimetric assay)
  • Simple "mix-and-measure" procedure allows reliable quantitation of creatine within 30 minutes
Creatinine is present in vertebrates and helps to supply energy to muscle
BioAssay Systems’ creatine assay is based on enzymatic reactions leading to formation of a pink colored product. The optical density at 570 nm or fluorescence intensity at λ ex/em = 530/585 nm is directly proportional to the creatine concentration in the sample.
In humans and animals, approximately half of creatine originates from food (mainly from fresh meat). Creatine supplementation has been investigated as a possible therapeutic approach for the treatment of muscular, neuromuscular, neurological and neurodegenerative diseases. Simple, direct and automation-ready. Procedures for measuring creatine are popular in research and drug discovery.
Customers Who Bought This Also Bought