ContactUsLinkComponent
EnzyChrom™ Catalase Assay Kit, BioAssay Systems
EnzyChrom™ Catalase Assay Kit, BioAssay Systems
# de catalogue 75878-056
Fournisseur:  BIOASSAY SYSTEMS
Numéro CAS:  
EnzyChrom* Catalase Assay Kit 100 tests
EnzyChrom™ Catalase Assay Kit, BioAssay Systems
# de catalogue 75878-056
Fournisseur:  BIOASSAY SYSTEMS
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Catalase Assay Kit
  • Size:
    100 assays
  • Method:
    OD570 nm, or FL530/585 nm
  • Sample:
    Serum, plasma, urine, saliva, cell culture etc.
  • Processing time:
    40 minutes
  • Cat. no.:
    75878-056
  • Supplier No.:
    ECAT-100
Spécifications
A propos de cet article
For quantitative determination of catalase activity and evaluation of drug effects on catalase activity.
  • Linear detection range 0.2 to 5 U/L catalase activity
Catalase (EC 1.11.1.6), is an ubiquitous antioxidant enzyme that catalyzes the decomposition of hydrogen peroxide (H2O2) to water and oxygen
BioAssay Systems’ improved assay directly measures catalase degradation of H2O2 using a redox dye. The change in color intensity at 570nm or fluorescence intensity (λex/em = 530/585nm) is directly proportional to the catalase activity in the sample.
By preventing excessive H2O2 build up, catalase allows important cellular processes which produce H2O2 as a byproduct to occur safely. Deficiency in catalase activity has been associated with grey hair and peroxisomal disorder acatalasia. Simple, direct and high-throughput assays for catalase activity find wide applications.
Customers Who Bought This Also Bought