ContactUsLinkComponent
EnzyChrom™ Glucose Assay Kit, BioAssay Systems
EnzyChrom™ Glucose Assay Kit, BioAssay Systems
# de catalogue 75878-052
Fournisseur:  BIOASSAY SYSTEMS
Numéro CAS:  
EnzyChrom* Glucose Assay Kit 100 tests
EnzyChrom™ Glucose Assay Kit, BioAssay Systems
# de catalogue 75878-052
Fournisseur:  BIOASSAY SYSTEMS
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Glucose Assay Kit
  • Size:
    100 assays
  • Method:
    OD570 nm, or FL530/585 nm
  • Sample:
    Serum, plasma, urine, saliva, milk, culture medium, food, agriculture etc.
  • Processing time:
    40 minutes
  • Cat. no.:
    75878-052
  • Supplier No.:
    EBGL-100
Spécifications
A propos de cet article
For quantitative colorimetric/fluorometric determination of glucose and evaluation of drug effects on glucose metabolism.
  • Linear detection range 5 to 300 μM (90 μg/dL to 5.4 mg/dL) glucose for colorimetric assays and 1 to 30 μM for fluorimetric assays
Glucose (C6H12O6) is a key diagnostic parameter for many metabolic disorders
BioAssay Systems’ glucose assay kit uses a single Working Reagent that combines the glucose oxidase reaction and color reaction in one step. The color intensity of the reaction product at 570 nm or fluorescence intensity at λex/em = 530/585 nm is directly proportional to glucose concentration in the sample.
Increased glucose levels have been associated with diabetes mellitus, hyperactivity of thyroid, pituitary and adrenal glands. Decreased levels are found in insulin secreting tumors, myxedema, hypopituitarism and hypoadrenalism. Simple, direct and high-throughput assays for measuring glucose concentrations find wide applications in research and drug discovery.
Customers Who Bought This Also Bought