ContactUsLinkComponent
EnzyLight™ ATP Assay Kit, BioAssay Systems
EnzyLight™ ATP Assay Kit, BioAssay Systems
# de catalogue 75878-050
Fournisseur:  BIOASSAY SYSTEMS
Numéro CAS:  
EnzyLight* ATP Assay Kit 100 tests
EnzyLight™ ATP Assay Kit, BioAssay Systems
# de catalogue 75878-050
Fournisseur:  BIOASSAY SYSTEMS
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    ATP Assay Kit
  • Size:
    100 assays
  • Method:
    Luminescence
  • Sample:
    Cells, tissue lysate
  • Processing time:
    10 minutes
  • Cat. no.:
    75878-050
  • Supplier No.:
    EATP-100
Spécifications
A propos de cet article
For rapid, quantitative, bioluminescent determination of ATP and evaluation of drug effects on ATP metabolism.
  • Safe. Non-radioactive assay. As low as 0.02 μM ATP or a single cell can be quantified
  • Convenient "mix-incubate-measure" type assay. No wash and reagent transfer steps are involved
  • Can be readily automated on HTS liquid handling systems
Adenosine 5’-triphosphate (ATP) is the chemical energy for cellular metabolism and is often referred to as “energy currency" of the cell
BioAssay Systems’ EnzyLight™ ATP Assay Kit provides a rapid method to measure intracellular ATP. The single working reagent lyses cells to release ATP, which, in the presence of luciferase, immediately reacts with the Substrate D-luciferin to produce light. The light intensity is a direct measure of intracellular ATP concentration. This non-radioactive, homogeneous cell-based assay is performed in microplates. The reagent is compatible with all liquid handling systems for high-throughput screening applications in 96-well and 384-well plates.
ATP is produced only in living cells during photosynthesis and cellular respiration and consumed in cellular processes including biosynthetic reactions, motility and cell division. It is a key indicator of cellular activity and has been utilized as a measure of cell viability and cytotoxicity in research and drug discovery.
Customers Who Bought This Also Bought