ContactUsLinkComponent
EnzyChrom™ Aspartate Transaminase Assay Kit, BioAssay Systems
EnzyChrom™ Aspartate Transaminase Assay Kit, BioAssay Systems
# de catalogue 75878-048
Fournisseur:  BIOASSAY SYSTEMS
Numéro CAS:  
EnzyChrom* Aspartate Transaminase Assay Kit 100 tests
EnzyChrom™ Aspartate Transaminase Assay Kit, BioAssay Systems
# de catalogue 75878-048
Fournisseur:  BIOASSAY SYSTEMS
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Aspartate Transaminase Assay Kit
  • Size:
    100 assays
  • Method:
    OD340 nm
  • Sample:
    Serum, plasma, etc.
  • Processing time:
    10 minutes
  • Cat. no.:
    75878-048
  • Supplier No.:
    EASTR-100
Spécifications
A propos de cet article
For quantitative determination of aspartate transaminase (AST) activity and evaluation of drug effects on AST activity.
  • Linear detection range 2-100 U/L
  • Assay can be carried out in a microplate or a cuvette and takes only 10 minutes
Aspartate Transaminase (AST), also known as serum glutamic oxaloacetic transaminase (GOT) or aspartate aminotransferase (ASAT/AAT), facilitates the conversion of aspartate and a-ketoglutarate to oxaloacetate and glutamate
BioAssay Systems’ AST activity assay is based on the quantification of oxaloacetate produced by AST. In this assay, oxaloacetate and NADH are converted to malate and NAD by the enzyme malate dehydrogenase. The decrease in NADH absorbance at 340 nm is proportionate to AST activity.
There are two isoenzymes in humans: GOT1 is a cytosolic isoenzyme derived from red blood cells and heart; GOT2 is the mitochondrial isoenzyme found mainly in the liver. AST is elevated in liver and muscle diseases. It is part of diagnostic tests for liver function, myocardial infarction, acute pancreatitis, acute hemolytic anemia, severe burns, acute renal disease and trauma. Simple, direct and automation-ready. Procedures for measuring AST activity find wide applications in research and drug discovery.
Customers Who Bought This Also Bought