Kevlar® Nomex® 2610 Seamless Heat Resistant Gloves, Wells Lamont
Fournisseur: Wells Lamont
Certificats
A propos de cet article
Kevlar® Nomex® heat resistant gloves protect up to 260 °C (500 °F).
Kevlar® Nomex® heat resistant gloves feature an outer shell that provides excellent thermal properties, allowing wearer to handle hot objects. These gloves are fully lined with cotton for softness. Gloves are ambidextrous, knit, and they feature a continuous cuff.
Ordering information: These gloves are sold by the dozen pieces.
Spécifications
- Catalog No:
- 75836-508
- 75836-510
- 75836-504
- 75836-506
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Latex/Latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Liner material:
- 100% Cotton
- 100% Cotton
- 100% Cotton
- 100% Cotton