ContactUsLinkComponent
EnzyChrom™ Aspartate Assay Kit, BioAssay Systems
EnzyChrom™ Aspartate Assay Kit, BioAssay Systems
# de catalogue 75878-046
Fournisseur:  BIOASSAY SYSTEMS
Numéro CAS:  
EnzyChrom* Aspartate Assay Kit 100 tests
EnzyChrom™ Aspartate Assay Kit, BioAssay Systems
# de catalogue 75878-046
Fournisseur:  BIOASSAY SYSTEMS
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Aspartate Assay Kit
  • Size:
    100 assays
  • Method:
    OD570 nm, or FL530/585 nm
  • Sample:
    Plasma, serum, tissue, culture media etc.
  • Processing time:
    60 minutes
  • Cat. no.:
    75878-046
  • Supplier No.:
    EASP-100
Spécifications
A propos de cet article
For quantitative determination of aspartate and evaluation of drug effects on aspartate metabolism.
  • Linear detection range 2 to 400 μM aspartate for colorimetric assays and 1 to 50 μM for fluorimetric assays
Aspartate, a nonessential amino acid, is a precursor to several other amino acids and is an excitatory neurotransmitter
BioAssay Systems’ Aspartate Assay Kit provides a simple, direct and automation-ready procedure for measuring aspartate concentration. Aspartate is converted into pyruvate which is then oxidized with the conversion of the dye into a colored and fluorescent form. The color intensity of the oxidized dye at 570 nm or fluorescence intensity at λex/em = 530/585 nm is directly proportional to the aspartate concentration in the sample.
Aspartate is involved in the urea cycle and gluconeogenesis.
Customers Who Bought This Also Bought