ContactUsLinkComponent
EnzyChrom™ Ascorbic Acid Assay Kit, BioAssay Systems
EnzyChrom™ Ascorbic Acid Assay Kit, BioAssay Systems
# de catalogue 75878-044
Fournisseur:  BIOASSAY SYSTEMS
Numéro CAS:  
EnzyChrom* Ascorbic Acid Assay Kit 100tests
EnzyChrom™ Ascorbic Acid Assay Kit, BioAssay Systems
# de catalogue 75878-044
Fournisseur:  BIOASSAY SYSTEMS
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Ascorbic Acid Assay Kit
  • Size:
    100 assays
  • Method:
    OD570 nm, or FL530/585 nm
  • Sample:
    Serum, plasma, urine, saliva, milk, tissue, and cell culture.
  • Processing time:
    10 minutes
  • Cat. no.:
    75878-044
  • Supplier No.:
    EASC-100
Spécifications
A propos de cet article
For quantitative determination of ascorbate (ascorbic acid) and evaluation of drug effects on ascorbic acid metabolism.
  • Linear detection range colorimetric assay 6 to 1000 μM, fluorimetric assay 1 to 100 μM ascorbic acid
Ascorbic acid (the L-enantiomer commonly known as vitamin C) is an important antioxidant found in living organisms and applied as additives in food and other industrial processes
BioAssay Systems’ method provides a simple, direct and high-throughput assay for measuring ascorbic acid. In this assay, ascorbic acid is oxidized by ascorbate oxidase resulting in the production of H2O2 which reacts with a specific dye to form a pink colored product. The color intensity at 570nm or fluorescence intensity (530/585 nm) is directly proportional to the ascorbic acid concentration in the sample.
By reacting with reactive oxygen species, it protects the cell from oxidative damages.
Customers Who Bought This Also Bought