ContactUsLinkComponent
EnzyChrom™ Adipolysis Assay Kit, BioAssay Systems
EnzyChrom™ Adipolysis Assay Kit, BioAssay Systems
# de catalogue 75878-042
Fournisseur:  BIOASSAY SYSTEMS
Numéro CAS:  
EnzyChrom* Adipolysis Assay Kit 200 tests
EnzyChrom™ Adipolysis Assay Kit, BioAssay Systems
# de catalogue 75878-042
Fournisseur:  BIOASSAY SYSTEMS
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Adipolysis Assay Kit
  • Size:
    200 assays
  • Method:
    OD570 nm, or FL530/585 nm
  • Sample:
    Cell culture media
  • Processing time:
    20 minutes
  • Cat. no.:
    75878-042
  • Supplier No.:
    EAPL-200
Spécifications
A propos de cet article
For quantitative assay of adipolysis and evaluation of drug effects on adipolysis.
  • Linear detection range 0.92 to 100 μg/mL (10 to 1000 μM) glycerol for colorimetric assays and 0.2 to 5 μg/mL for fluorimetric assays
  • Robust and amenable to HTS assays
Obesity is a chronic condition that develops from storage of excessive energy in the form of adipose tissue
BioAssay Systems’ adipolysis assay kit directly measures glycerol released during adipolysis. This homogeneous assay uses a single Working Reagent that combines glycerol kinase, glycerol phosphate oxidase and color reactions in one step. The color intensity of the reaction product at 570 nm is directly proportional to glycerol concentration in the sample.
The resulting adiposity presents a high risk factor for diseases such as type 2 diabetes, cardiovascular diseases, and cancer. ADIPOLYSIS or lipolysis is a highly regulated process in fat metabolism, in which triglycerides are broken down into glycerol and free fatty acids. Rapid, robust and accurate. Procedures for adipolysis quantification in cell culture are very useful in research and drug discovery.
Customers Who Bought This Also Bought