ContactUsLinkComponent
EnzyChrom™ Alanine Assay Kit, BioAssay Systems
EnzyChrom™ Alanine Assay Kit, BioAssay Systems
# de catalogue 75878-036
Fournisseur:  BIOASSAY SYSTEMS
Numéro CAS:  
EnzyChrom* L-Alanine Assay Kit 100 tests
EnzyChrom™ Alanine Assay Kit, BioAssay Systems
# de catalogue 75878-036
Fournisseur:  BIOASSAY SYSTEMS
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Alanine Assay Kit
  • Size:
    100 assays
  • Method:
    OD570 nm, or FL530/585 nm
  • Sample:
    Plasma, serum, urine, tissue and culture media.
  • Processing time:
    60 minutes
  • Cat. no.:
    75878-036
  • Supplier No.:
    EALA-100
Spécifications
A propos de cet article
For quantitative determination of L-Alanine and evaluation of drug effects on alanine metabolism.
  • Linear detection range 1 to 200 μM alanine for colorimetric assays and 0.4 to 20 μM for fluorimetric assays
Alanine, a nonessential amino acid, is utilized in the glucose-alanine cycle between tissues and the liver
BioAssay Systems’ Alanine Assay Kit provides a simple, direct and automation-ready procedure for measuring alanine concentration. Alanine is converted into pyruvate which can then be directly measured. The color intensity of the reaction product at 570nm or fluorescence intensity at λex/em = 530/585nm is directly proportional to the alanine concentration in the sample.
In tissues that metabolize amino acids, amino groups are collected as glutamate by transamination. The amine group is then transferred by alanine transaminase (ALT) from glutamate to pyruvate to form alanine and a-ketoglutarate. The alanine generated is transported to the liver where a reverse ALT reaction occurs and pyruvate is regenerated. Pyruvate is converted through gluconeogenesis to glucose which can then be recirculated to the tissues. Alanine concentration may have some correlation with high blood pressure, energy intake, cholesterol levels and body mass index.
Customers Who Bought This Also Bought