ContactUsLinkComponent
EnzyLight™ ADP Assay Kit, BioAssay Systems
EnzyLight™ ADP Assay Kit, BioAssay Systems
# de catalogue 75878-034
Fournisseur:  BIOASSAY SYSTEMS
Numéro CAS:  
EnzyLight* ADP Assay Kit 100 tests
EnzyLight™ ADP Assay Kit, BioAssay Systems
# de catalogue 75878-034
Fournisseur:  BIOASSAY SYSTEMS
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    ADP Assay Kit
  • Size:
    100 assays
  • Method:
    Luminescence
  • Sample:
    Cells
  • Processing time:
    20 minutes
  • Cat. no.:
    75878-034
  • Supplier No.:
    EADP-100
Spécifications
A propos de cet article
For rapid, quantitative, bioluminescent determination of ADP concentration and evaluation of drug effects on ADP metabolism.
  • Safe
  • Non-radioactive assay
  • As low as 0.02 μM ADP can be quantified
  • Can be readily automated to process thousands of samples per day
BioAssay Systems’ EnzyLight™ ADP Assay Kit provides a rapid method to measure ADP levels. The assay involves two steps. In the first step, the working reagent lyses cells to release ATP and ADP. In the presence of luciferase, ATP immediately reacts with the Substrate D-luciferin to produce light. The light intensity is a direct measure of intracellular ATP concentration. In the second step, the ADP is converted to ATP through an enzyme reaction. This newly formed ATP then reacts with the D-luciferin as in the first step. The second light intensity measured represents the total ADP and ATP concentration in the sample. This non-radioactive, homogeneous cell-based assay is performed in microplates. The reagent is compatible with all culture media and with all liquid handling systems for high-throughput screening applications in 96-well and 384-well plates.
Customers Who Bought This Also Bought