ContactUsLinkComponent
EnzyChrom™ Acetylcholine Assay Kit, BioAssay Systems
EnzyChrom™ Acetylcholine Assay Kit, BioAssay Systems
# de catalogue 75878-030
Fournisseur:  BIOASSAY SYSTEMS
Numéro CAS:  
EnzyChrom* Acetylcholine Assay Kit 100tests
EnzyChrom™ Acetylcholine Assay Kit, BioAssay Systems
# de catalogue 75878-030
Fournisseur:  BIOASSAY SYSTEMS
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Acetylcholine Assay Kit
  • Size:
    100 assays
  • Method:
    OD570 nm, or FL530/585 nm
  • Sample:
    Serum, plasma, urine, saliva, milk, tissue, and cell culture etc.
  • Processing time:
    30 minutes
  • Cat. no.:
    75878-030
  • Supplier No.:
    EACL-100
Spécifications
A propos de cet article
For quantitative determination of acetylcholine and evaluation of drug effects on acetylcholine metabolism.
  • Linear detection range colorimetric assay 10 to 200 μM, fluorimetric assay 0.4 to 10 μM acetylcholine
Acetylcholine is a neurotransmitter produced in acetylcholinergic neurons. It plays important roles in skeletal muscle movement, regulation of smooth and cardiac muscles, as well as in learning, memory and mood. BioAssay Systems’ method provides a simple, direct and high-throughput assay for measuring acetylcholine in biological samples. In this assay, acetylcholine is hydrolyzed by acetylcholinesterase to choline which is oxidized by choline oxidase to betaine and H2O2. The resulting H2O2 reacts with a specific dye to form a pink colored product. The color intensity at 570nm or fluorescence intensity (530/585 nm) is directly proportional to the acetylcholine concentration in the sample.
Customers Who Bought This Also Bought