ContactUsLinkComponent
EnzyChrom™ Starch Assay Kit, BioAssay Systems
EnzyChrom™ Starch Assay Kit, BioAssay Systems
# de catalogue 75878-028
Fournisseur:  BIOASSAY SYSTEMS
Numéro CAS:  
EnzyChrom* Starch Assay Kit 100 tests
EnzyChrom™ Starch Assay Kit, BioAssay Systems
# de catalogue 75878-028
Fournisseur:  BIOASSAY SYSTEMS
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Starch Assay Kit
  • Size:
    100 assays
  • Method:
    OD570 nm, or FL530/585 nm
  • Sample:
    Biological, agriculture, food etc.
  • Processing time:
    30 minutes
  • Cat. no.:
    75878-028
  • Supplier No.:
    E2ST-100
Spécifications
A propos de cet article
For quantitative determination of starch and evaluation of drug effects on starch metabolism.
  • Linear detection range 2 - 200 μg/mL starch for colorimetric assays and 0.2 - 20 μg/mL for fluorimetric assays
Starch, chemical formula (C6H10O5)n, is a polysaccharide carbohydrate consisting of a large number of glucose units joined together by glycosidic bonds
BioAssay Systems’ starch uses a single Working Reagent that combines the enzymatic break down of starch and the detection of glucose in one step. The color intensity of the reaction product at 570 nm or fluorescence intensity at λex/em = 530/585 nm is directly proportional to the starch concentration in the sample. This simple convenient assay is carried out at room temperature and takes only 30 min.
All plant seeds and tubers contain starch present in the form of amylose and amylopectin. Starch is the most consumed polysaccharide in the human diet. Some starches are digested very quickly, and cause a rapid and large rise in blood sugar. Others are digested more slowly, and some starch, called resistant starch, is not digested in the small intestine at all, and thus causes little or no blood sugar rise. Simple, direct and automation-ready. Procedures for measuring starch concentrations find wide applications in research and drug discovery.
Customers Who Bought This Also Bought