ContactUsLinkComponent
EnzyChrom™ Glycogen Assay Kit, BioAssay Systems
EnzyChrom™ Glycogen Assay Kit, BioAssay Systems
# de catalogue 75878-022
Fournisseur:  BIOASSAY SYSTEMS
Numéro CAS:  
EnzyChrom* Glycogen Assay Kit 100 tests
EnzyChrom™ Glycogen Assay Kit, BioAssay Systems
# de catalogue 75878-022
Fournisseur:  BIOASSAY SYSTEMS
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Glycogen Assay Kit
  • Size:
    100 assays
  • Method:
    OD570 nm, or FL530/585 nm
  • Sample:
    Biological
  • Processing time:
    30 minutes
  • Cat. no.:
    75878-022
  • Supplier No.:
    E2GN-100
Spécifications
A propos de cet article
For quantitative determination of glycogen and evaluation of drug effects on glycogen metabolism.
  • Linear detection range 2 to 200 μg/mL glycogen for colorimetric assays and 0.2 to 20 μg/mL for fluorimetric assays
Glycogen is a branched polysaccharide of glucose units linked by α-1,4 glycosidic bonds and α-1,6 glycosidic bonds
BioAssay Systems’ glycogen assay uses a single Working Reagent that combines the enzymatic break down of glycogen and the detection of glucose in one step. The color intensity of the reaction product at 570nm or fluorescence intensity at λex/em = 530/585 nm is directly proportional to the glycogen concentration in the sample. This simple convenient assay is carried out at room temperature and takes only 30 min.
It is stored primarily in the liver and muscle, and forms an energy reserve that can be quickly mobilized to meet a sudden need for glucose. The most common glycogen metabolism disorder is found in diabetes, in which, due to abnormal amounts of insulin, liver glycogen can be abnormally accumulated or depleted. Genetic glycogen storage diseases have been associated with various inborn errors of metabolism caused by deficiencies of enzymes necessary for glycogen synthesis or breakdown. Simple, direct and automation-ready. Procedures for measuring glycogen concentrations find wide applications in research and drug discovery.
Customers Who Bought This Also Bought