ContactUsLinkComponent
EnzyChrom™ ADP Assay Kit, BioAssay Systems
EnzyChrom™ ADP Assay Kit, BioAssay Systems
# de catalogue 75878-018
Fournisseur:  BIOASSAY SYSTEMS
Numéro CAS:  
EnzyChrom* ADP Assay Kit 100 tests
EnzyChrom™ ADP Assay Kit, BioAssay Systems
# de catalogue 75878-018
Fournisseur:  BIOASSAY SYSTEMS
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    ADP Assay Kit
  • Size:
    100 assays
  • Method:
    FL530/590 nm
  • Sample:
    Cells and other biological samples
  • Processing time:
    30 minutes
  • Cat. no.:
    75878-018
  • Supplier No.:
    E2ADP-100
Spécifications
A propos de cet article
For quantitative fluorometric determination of ADP determination in cells and other biological samples.
  • Safe and sensitive
  • Non-radioactive assay
  • As low as 0.1 μM ADP can be quantified
  • Can be readily automated to process thousands of samples per day
Adenosine diphosphate (ADP) is the product of ATP dephosphorylation by ATPases
BioAssay Systems’ newly designed ADP Assay Kit provides a convenient fluorometric means to measure ADP level even in the presence of ATP. In the assay, ADP is converted to ATP and pyruvate. The generated pyruvate is then quantified by a fluorimetric method (λex/em = 530/590nm). The assay is simple, sensitive, stable, high-throughput adaptable and can detect as low as 0.1 μM ADP in biological samples.
ADP can be converted back to ATP by ATP synthases. ADP levels regulate several enzymes involved in intermediary metabolism. Conventionally, ADP levels are measured by luciferase/luciferin mediated assays after ADP is converted to ATP. However, since these assays require measurement of ATP in the sample before conversion of ADP to ATP, if the nascent ATP concentration is significantly higher than the ADP concentration, the ATP signal will drown out the ADP signal.
Customers Who Bought This Also Bought