Spécifications
- Description:ADP Assay Kit
- Size:100 assays
- Method:FL530/590 nm
- Sample:Cells and other biological samples
- Processing time:30 minutes
- Cat. no.:75878-018
- Supplier No.:E2ADP-100
Spécifications
A propos de cet article
For quantitative fluorometric determination of ADP determination in cells and other biological samples.
- Safe and sensitive
- Non-radioactive assay
- As low as 0.1 μM ADP can be quantified
- Can be readily automated to process thousands of samples per day
Adenosine diphosphate (ADP) is the product of ATP dephosphorylation by ATPases
BioAssay Systems’ newly designed ADP Assay Kit provides a convenient fluorometric means to measure ADP level even in the presence of ATP. In the assay, ADP is converted to ATP and pyruvate. The generated pyruvate is then quantified by a fluorimetric method (λex/em = 530/590nm). The assay is simple, sensitive, stable, high-throughput adaptable and can detect as low as 0.1 μM ADP in biological samples.
ADP can be converted back to ATP by ATP synthases. ADP levels regulate several enzymes involved in intermediary metabolism. Conventionally, ADP levels are measured by luciferase/luciferin mediated assays after ADP is converted to ATP. However, since these assays require measurement of ATP in the sample before conversion of ADP to ATP, if the nascent ATP concentration is significantly higher than the ADP concentration, the ATP signal will drown out the ADP signal.