Spinner Flask, Water Jacketed, Bioprocess, Chemglass
Fournisseur: Chemglass
Certificats
A propos de cet article
Water jacketed, flat bottomed spinner flasks consist of a water jacketed spinner flask and a microcarrier style impeller with glass shaft that extends through the cap and PTFE paddle.
- Manufactured from Type 1 Class A borosilicate glass
Flasks are typically used with a circulating water bath. Jacket volumes are approximate and flasks are not graduated.
Certifications: Glass conforms to ASTM Type 1, Class A requirements.
BodyTextAccessories: Port assemblies are constructed of 316 l electro-polished stainless steel. These port assemblies fit all types of spinner flasks. The assemblies consist of a siphon and vent tube. They are supplied complete with a GL-45 open top cap and silicone gasket.
Customers Who Bought This Also Bought