ContactUsLinkComponent
Uric Acid Assay Kit, BioAssay Systems
Uric Acid Assay Kit, BioAssay Systems
# de catalogue 75878-102
Fournisseur:  BIOASSAY SYSTEMS
Numéro CAS:  
QuantiChrom* Uric Acid Assay Kit 250tests
Uric Acid Assay Kit, BioAssay Systems
# de catalogue 75878-102
Fournisseur:  BIOASSAY SYSTEMS
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Uric Acid Assay Kit
  • Size:
    250 assays
  • Method:
    OD590 nm
  • Sample:
    Serum, plasma, urine and other biological samples.
  • Processing time:
    30 minutes
  • Brand name:
    QuantiChrom™
  • Cat. no.:
    75878-102
  • Supplier No.:
    DIUA-250
Spécifications
A propos de cet article
For quantitative determination of uric acid and evaluation of drug effects on uric acid metabolism
  • Sensitive and accurate. Use 5 μL samples. Linear detection range 0.22 mg/dL (13 μM) to 30 mg/dL (1785 μM) uric acid in 96-well plate assay.
  • Simple and high-throughput. The procedure involves addition of a single working reagent and incubation for 30 min. Can be readily automated as a high-throughput assay in 96-well plates for thousands of samples per day.
  • Improved reagent stability and versatility. The optimized formulation has greatly enhanced reagent and signal stability. Cuvet or 96-well plate assay.
  • Low interference in biological samples. No pretreatments are needed. Assays can be directly performed on serum samples.
  • Method: OD590nm
  • Samples: Serum, plasma, urine and other biological samples
Uric acid is the waste product produced from the degradation of purines. In healthy human, uric acid is filtered and removed from the blood by the kidneys and excreted into urine. Because a number of kidney diseases are known to affect uric acid levels, uric acid determination is thus important and useful in diagnosing and evaluating kidney diseases. For example, when uric acid is present in the blood at abnormally high levels, it tends to crystallize in body joints, resulting in gout, a very painful inflammatory condition. Increased levels of uric acid are also known to be associated with uremia, leukemia and pneumonia. Simple, direct and automation-ready procedures for measuring uric acid concentration in blood are becoming popular in Research and Drug Discovery. BioAssay Systems’ uric acid assay kit is designed to measure uric acid directly in serum without any pretreatment. The improved method utilizes 2,4,6-tripyridyl-s-triazine that forms a blue colored complex specifically with iron in the presence of uric acid. The intensity of the color, measured at 590nm, is directly proportional to the uric acid concentration in the serum. The optimized formulation substantially reduces interference by substances in the raw samples.
Customers Who Bought This Also Bought