ContactUsLinkComponent
QuantiChrom™ Peroxide Assay Kit, BioAssay Systems
QuantiChrom™ Peroxide Assay Kit, BioAssay Systems
# de catalogue 75878-002
Fournisseur:  BIOASSAY SYSTEMS
Numéro CAS:  
QuantiChrom* Peroxide Assay Kit 250 tests
QuantiChrom™ Peroxide Assay Kit, BioAssay Systems
# de catalogue 75878-002
Fournisseur:  BIOASSAY SYSTEMS
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Peroxide Assay Kit
  • Size:
    250 assays
  • Method:
    OD585 nm
  • Sample:
    Serum, citrate-plasma, urine, cell lysate, culture media etc
  • Processing time:
    30 minutes
  • Cat. no.:
    75878-002
  • Supplier No.:
    DIOX-250
Spécifications
A propos de cet article
For quantitative determination of peroxide and evaluation of drug effects on peroxide metabolism.
  • Enhanced color intensity using sorbitol
  • Detection range 0.2 - 30 μM (7 - 1,020 ng/mL) H2O2 in 96-well plate assay
  • Can be readily automated to process thousands of samples per day
Peroxide (e.g
BioAssay Systems’ peroxide assay kit is designed to measure peroxide concentration in biological samples without any pretreatment. The improved method utilizes the chromogenic Fe3+-xylenol orange reaction, in which a purple complex is formed when Fe2+ provided in the reagent is oxidized to Fe3+ by peroxides present in the sample. The intensity of the color, measured at 540-610nm, is an accurate measure of the peroxide level in the sample. The optimized formulation substantially reduces interference by substances in the raw samples.
hydrogen peroxide H2O2) is one of the key reactive oxygen species formed under oxidative stress conditions. High levels of peroxide formation have been linked to pathological conditions such as ageing, asthma, diabetes, atherosclerosis, cataract, inflammatory arthritis and neurodegenerative diseases. Simple, direct and automation-ready. Procedures for quantitative determination of peroxide find wide applications in research and drug discovery.
Customers Who Bought This Also Bought