Learning Resources Attribute Blocks, Set of 60
# de catalogue 470002-850
Fournisseur: PRECISION MOULDED POLYMERS LTD
Spécifications
- Description:Attribute Blocks, Set of 60
- Height:1.4"
- Cat. no.:470002-850
- Length:8.5"
- Width:7.2"
- Supplier No.:1001
Spécifications
A propos de cet article
Teach sorting, classification, and size comparison.
- Includes tracing template and a storage tray with a shape sorter lid
- Features five shapes in two sizes, two thicknesses, and three colors
Teach sorting, patterns, size comparison and other early math skills with these durable plastic blocks. This 60-piece set includes five different shapes in two sizes, two thicknesses, and three colors. Sets come in a handy storage tray with shape sorter that can also be used for tracing. Activity Guide included.
Caution: Includes tracing template and a storage tray with a shape sorter lid
Features five shapes in two sizes, two thicknesses, and three colors
Customers Who Bought This Also Bought