ContactUsLinkComponent
QuantiChrom™ Chloride Assay Kit, BioAssay Systems
QuantiChrom™ Chloride Assay Kit, BioAssay Systems
# de catalogue 75877-988
Fournisseur:  BIOASSAY SYSTEMS
Numéro CAS:  
QuantiChrom* Chloride Assay Kit 250 tests
QuantiChrom™ Chloride Assay Kit, BioAssay Systems
# de catalogue 75877-988
Fournisseur:  BIOASSAY SYSTEMS
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Chloride Assay Kit
  • Size:
    250 assays
  • Method:
    OD610 nm
  • Sample:
    Biological, food and environment.
  • Cat. no.:
    75877-988
  • Supplier No.:
    DICL-250
Spécifications
A propos de cet article
For quantitative determination of chloride ion and evaluation of drug effects on chloride metabolism.
  • Linear detection range 0.7 mg/dL (0.2 mM) to 35 mg/dL (10 mM) chloride ion
  • The procedure involves addition of a single working reagent and incubation for five minutes
  • Can readily automated to process thousands of samples per day
  • Assays can be executed in microplate or cuvet
  • Low interference in biological samples
Chloride is the major extracellular anion in human body fluids
BioAssay Systems’ chloride assay kit is designed to measure chloride directly in biological samples without any pretreatment. The improved Fried method utilizes mercuric 2,4,6-tripyridyl-s-triazine, which forms a colored complex specifically with chloride. The intensity of the color, measured at 610nm, is directly proportional to the chloride concentration in the sample. The optimized formulation substantially reduces interference by substances in the raw samples.
Chloride plays a key role in maintaining proper water distribution, osmotic pressure and electrolyte balance. Low chloride concentrations may be found with prolonged vomiting, extensive burns, metabolic acidosis, Addisonia crisis and renal diseases. Elevated chloride concentrations are associated with dehydration, congestive heart failure, hyperventilation and urinary obstructions. Determination of chloride in sweat is useful in diagnosing cystic fibrosis. Simple, direct and automation-ready. Procedures for measuring chloride concentration in biological samples are becoming popular in Research and Drug Discovery.
Customers Who Bought This Also Bought