Spécifications
- Description:Calcium Assay Kit
- Size:500 assays
- Method:OD612nm
- Sample:Biological, food, environment
- Processing time:Three minutes
- Cat. no.:75877-986
- Supplier No.:DICA-500
Spécifications
A propos de cet article
For quantitative determination of calcium ion and evaluation of drug effects on calcium metabolism.
- Linear detection range 0.08 - 20 mg/dL (20 - 5000 μM) calcium ions
- Can readily automated to process thousands of samples per day
- Assays can be executed in microplate or cuvet
Calcium is measured to monitor diseases of the bone or calcium regulation disorders
BioAssay Systems’ calcium assay kit is designed to measure calcium directly in biological samples without any pretreatment. A phenolsulphonephthalein dye in the kit forms a very stable blue colored complex specifically with free calcium. The intensity of the color, measured at 612 nm, is directly proportional to the calcium concentration in the sample. The optimized formulation minimizes any interference by substances such as magnesium, lipid, protein and bilirubin.
Increased calcium levels in serum are reported in hyperparathyroidism, metastatic bone lesions and hypervitaminosis, while decreased levels are observed in hypoparathyroidism, nephrosis, rickets, steatorrhea, nephritis and calcium-losing syndromes. Urinary calcium levels aid the clinician in understanding how the kidneys handle calcium in certain diseases of the parathyroid gland. Urinary calcium levels are also essential in the medical evaluation of kidney stones. Simple, direct and automation-ready. Procedures for measuring calcium concentration in biological samples are becoming popular in Research and Drug Discovery.