ContactUsLinkComponent
QuantiFluo™ Caspase Assay Kit, BioAssay Systems
QuantiFluo™ Caspase Assay Kit, BioAssay Systems
# de catalogue 75877-968
Fournisseur:  BIOASSAY SYSTEMS
Numéro CAS:  
QuantiFluo* Caspase-3 Assay Kit 100 tests
QuantiFluo™ Caspase Assay Kit, BioAssay Systems
# de catalogue 75877-968
Fournisseur:  BIOASSAY SYSTEMS
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Caspase Assay Kit
  • Size:
    100 assays
  • Method:
    FL400/490 nm
  • Sample:
    Cell and tissue lysate
  • Processing time:
    60 minutes
  • Cat. no.:
    75877-968
  • Supplier No.:
    DCS3-100
Spécifications
A propos de cet article
For quantitative determination of the apoptosis target caspase-3 activity and HTS screen for apoptosis inducers and inhibitors.
  • Safe
  • Non-radioactive assay
  • This homogeneous "mix-incubate-measure" type assay can be readily automated for processing thousands of samples per day
Caspases are members of the aspartate-specific cysteinyl protease family that play a central role in apoptosis
BioAssay Systems’ Caspase-3 Assay Kit provides a convenient means to measure caspase-3 activity in biological samples. In the assay, a specific substrate (N-Ac-DEVD-AFC) is cleaved by active caspase-3, forming a highly fluorescent product. The fluorescence intensity (λex/em = 400/490nm) is proportional to the caspase-3 activity.
Apoptosis is involved in a variety of physiological and pathological events, ranging from normal fetal development to diseases such as cancer, organ failure, and neurodegenerative diseases. Caspase-3 is key biomarker in the assessment of apoptosis and in understanding mechanism of apoptosis induction.
Customers Who Bought This Also Bought