ContactUsLinkComponent
QuantiChrom™ Chromium Assay Kit, BioAssay Systems
QuantiChrom™ Chromium Assay Kit, BioAssay Systems
# de catalogue 75877-966
Fournisseur:  BIOASSAY SYSTEMS
Numéro CAS:  
QuantiChrom* Chromium Assay Kit 250 tests
QuantiChrom™ Chromium Assay Kit, BioAssay Systems
# de catalogue 75877-966
Fournisseur:  BIOASSAY SYSTEMS
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Chromium Assay Kit
  • Size:
    250 assays
  • Method:
    OD480 nm
  • Sample:
    Biological (serum, plasma etc), environmental (water, soil etc), food and beverage samples.
  • Processing time:
    20 minutes
  • Cat. no.:
    75877-966
  • Supplier No.:
    DCRM-250
Spécifications
A propos de cet article
For quantitative determination of chromium Cr(VI).
  • Linear detection range of 20 - 2000 μg/L Chromium
  • Homogeneous "mix-incubate-measure" type assay
  • No wash and reagent transfer steps are involved
  • Can be readily automated for processing thousands of samples per day
Chromium is widely used in various industries such as electroplating, leather tanning, chrome paint, dying, hardened steel, ceramic and glass industry
BioAssay Systems’ Chromium Assay Kit provides a simple one-step colorimetric means to directly measure Cr(VI) in a sample. In the assay, Cr(VI) forms a stable complex with a specific chromogenic dye. The optical density at 480nm is directly proportionate to the Cr(VI) concentration in the sample. Cr(III) can be converted to Cr(VI) with nitric acid/hydrochloric acid, thus allowing the determination of Cr(III) or total Cr [Cr(III) + Cr(VI)] in the sample. The assay is sensitive with a detection limit of 20 μg/L Cr.
Chromium exists in two stable oxidation states, hexavalent Cr(VI) and trivalent Cr(III). Cr(VI) is produced solely by industrial processes, whereas in nature, chromium exists in its trivalent form. Cr(III) is generally regarded as nontoxic due to poor absorption. Cr(VI) is considered a pulmonary carcinogen and has tested positive in genotoxicity tests. It is one of the most serious pollutants in many water streams due to its carcinogenic potential. Most countries apply a legal limit of 50-100 μg/L Cr in drinking water.
Customers Who Bought This Also Bought