ContactUsLinkComponent
QuantiChrom™ Nitric Oxide Assay Kit, BioAssay Systems
QuantiChrom™ Nitric Oxide Assay Kit, BioAssay Systems
# de catalogue 75877-946
Fournisseur:  BIOASSAY SYSTEMS
Numéro CAS:  
QuantiChrom* Nitric Oxide Assay Kit 100 tests
QuantiChrom™ Nitric Oxide Assay Kit, BioAssay Systems
# de catalogue 75877-946
Fournisseur:  BIOASSAY SYSTEMS
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Nitric Oxide Assay Kit
  • Size:
    100 assays
  • Method:
    OD540nm
  • Sample:
    Plasma, serum, urine, tissue/cells and foods.
  • Processing time:
    40 minutes
  • Cat. no.:
    75877-946
  • Supplier No.:
    D2NO-100
Spécifications
A propos de cet article
For quantitative determination of nitric oxide and evaluation of drug effects on its metabolism.
  • Detection range 0.6 - 200 μM in 96-well plate
  • Rapid and reliable
  • Can be readily automated to measure thousands of samples per day
Nitric oxide (NO) is a reactive radical that plays an important role in many key physiological functions
QuantiChrom™ Nitric Oxide Assay Kit is designed to accurately measure NO production following reduction of nitrate to nitrite using improved Griess method. The procedure is simple and the time required for sample pretreatment and assay is reduced to as short as 30 min.
NO, an oxidation product of arginine by nitric oxide synthase, is involved in host defense and development, activation of regulatory proteins and direct covalent interaction with functional biomolecules. Simple, direct and automation-ready. Procedures for measuring NO are becoming popular in Research and Drug Discovery. Since NO is oxidized to nitrite and nitrate, it is common practice to quantitate total NO2-/NO3- as a measure for NO level. BioAssay Systems.
Customers Who Bought This Also Bought