ContactUsLinkComponent
Cell Viability and Proliferation Reagent, Biotium
Cell Viability and Proliferation Reagent, Biotium
# de catalogue 89138-264
Fournisseur:  Biotium
Numéro CAS:  
Cell Viability and Proliferation Reagent, Biotium
# de catalogue 89138-264
Fournisseur:  Biotium
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    XTT reagent
  • Size:
    100 mg
  • Cat. no.:
    89138-264
  • Supplier No.:
    10060
Spécifications
A propos de cet article
XTT reagent is a tetrazolium derivative used widely in cell viability and proliferation testing.
  • Pale yellow solid soluble in water
  • Store desiccated at 4°C
XTT reagent is a tetrazolium derivative used widely in cell viability and proliferation testing. This is readily reduced in viable cells to a highly water-soluble orange colored product. Applications of the reagent include studies of antifungal susceptibility, drug sensitivity of cells, parasitic nematode viability and tumor cell cytotoxicity.
Customers Who Bought This Also Bought