PTFE Tape, Chemglass
Catalog # 16002-900
Supplier: Chemglass
Small Business Enterprise
Specifications
- Width:1.9 cm (¾")
- Description:PTFE Tape
- Length:13.2 m (43.3')
- Cat. no.:16002-900
- Supplier No.:CG-1963-B-39
Specifications
About this item
PTFE tape may be used to create a leak-tight seal.
Customers Who Bought This Also Bought