NMR Tube Cleaner, 5-Position, CHEMGLASS INC.
Fournisseur: Chemglass
Certificats
A propos de cet article
This device enables the cleaning of five NMR tubes at the same time.
- Cleaning solvent is added into the adapter followed by air drying to complete the process in seconds
- Flexible tubing and adapter design reduces breakage
- Stopcock permits easy draining of solvent
- Hose connection is 10 mm at the largest serration
The NMR tube is placed over the tubing and seats in the adapter, leaving a small space between the end of the PTFE tubing and the inside bottom of the NMR tube. With the stopcock in the closed position, the reservoir is connected to a low vacuum source.
Customers Who Bought This Also Bought