PRO External Circulators with Base Controller, Lauda Brinkmann
Fournisseur: Lauda-Brinkmann
Certificats
A propos de cet article
Our PRO units have advantages such as low filling volume combined with powerful Varioflex pump, resulting in rapid temperature change. Environment Friendly (GWP).
- Bright high contrast OLED display
- Programmer with 1 program, 20 segments
- Removable remote control
- Temperature display in °C or °F
- Menu navigation in 5 languages
LAUDA PRO External Circulators are designed to handle the most demanding external applications
The BASE Controller features a bright OLED display that is removable from the unit, allowing for flexible operation.
The low internal filling volume of the PRO permits for rapid temperature changes and dynamic control of the external process. Varying temperature ranges and heating/cooling powers ensure the optimal fit.
Certifications: CE marked.
Customers Who Bought This Also Bought