PRO Internal Baths with Base Controller, Lauda-Brinkmann
Fournisseur: Lauda-Brinkmann
Certificats
A propos de cet article
PRO units have advantages such as low filling volume combined with powerful Varioflex pump, resulting in rapid temperature change. Environment Friendly (GWP).
- Bright high contrast OLED display
- Programmer with 1 program, 20 segments
- Removable remote control
- Temperature display in °C or °F
- Menu navigation in 5 languages
LAUDA PRO Thermostats are optimized for interal bath applications and offer excellent temperature stability
The BASE Controller features a bright OLED display that is removable from the unit, allowing for flexible operation.
A multistage adjustable pump ensures good mixing of the bath, ensuring a high level of temperature homogeneity. Varying sizes of baths and temperature ranges ensure the proper fit to each individual application.
Certifications: CE marked.
Customers Who Bought This Also Bought