SteriPlast® Sterile Scraper, Bürkle
# de catalogue 75876-072
Fournisseur: BURKLE INC
Spécifications
- Dimensions:200L x 80W mm (3.15 x 7.87")
- Description:SteriPlast® Sterile Scraper
- Cat. no.:75876-072
- Supplier No.:5378-1030
Spécifications
A propos de cet article
The SteriPlast® Sterile Scraper, made of polystyrene, is produced in a class 7 cleanroom (10,000), individually packaged for disposable use, and then sterilized by gamma rays.
- Polystyrene construction
- Individually packaged
- Sterilized by gamma rays
Sturdy and convenient scraper for disposable use. The scraper is suitable for scraping off, smoothing, or cleaning, as well as removing residue.
Packaging: 10 scrapers per package.
Customers Who Bought This Also Bought