SteriPlast® Scraper for Foodstuffs, Bürkle
# de catalogue 75876-068
Fournisseur: BURKLE INC
Spécifications
- Dimensions:200L x 80W mm (3.15 x 7.87")
- Color:Blue
- Description:SteriPlast® Scraper for Foodstuff
- Cat. no.:75876-068
- Supplier No.:5378-3030
Spécifications
A propos de cet article
The SteriPlast® Scraper for Foodstuffs, made of blue polystyrene, is produced in a class 7 cleanroom (10,000), individually packaged for disposable use, and then sterilized by gamma rays.
- Can be used as part of HACCP/IFS foreign object management
- Ideally suited for the food, animal feed, and pharmaceutical industries
Sturdy and convenient scraper for disposable use. The scraper is suitable for scraping off, smoothing, or cleaning, as well as removing residue.
Packaging: 10 scrapers per package.
Customers Who Bought This Also Bought