Avantor® Alltima HP C18 Hi-Load, HPLC Columns
Fournisseur: HICHROM LIMITED
Certificats
A propos de cet article
Avantor® Alltima HP columns combine a range of different phase chemistries with high purity silica.
- Excellent column stability
- Low to no detectable column bleed
- Multiple selectivity options
These columns have highest carbon load for superior retention and loadability for high resolution of complex samples.
These product family combines the selectivity and performance needed to overcome the most challenging separation needs. The low column bleed makes these columns ideal for microbore applications.
Column hardware: Stainless steel.
Spécifications
- Catalog No:
- 75797-504
- 75830-818
- 75797-502
- 75797-526
- 75797-532
- 75797-488
- 75797-530
- 75797-486
- 75797-492
- 75797-534
- 75797-490
- 75797-494
- 75797-472
- 75797-476
- 75797-474
- 75797-480
- 75797-478
- 75830-806
- 75797-520
- 75797-524
- 75797-506
- 75797-484
- 75797-482
- 75797-510
- 75797-508
- 75798-908
- 75797-514
- 75798-910
- 75797-512
- 75797-518
- 76398-380
- 75798-912
- Carbon load:
- 24%
- 24%
- 24%
- 24%
- 24%
- 24%
- 24%
- 24%
- 24%
- 24%
- 24%
- 24%
- 24%
- 24%
- 24%
- 24%
- 24%
- 24%
- 24%
- 24%
- 24%
- 24%
- 24%
- 24%
- 24%
- 24%
- 24%
- 24%
- 24%
- 24%
- 24%
- 24%
- Endcapped:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Surface area:
- 450 m²/g
- 450 m²/g
- 450 m²/g
- 450 m²/g
- 450 m²/g
- 450 m²/g
- 450 m²/g
- 450 m²/g
- 450 m²/g
- 450 m²/g
- 450 m²/g
- 450 m²/g
- 450 m²/g
- 450 m²/g
- 450 m²/g
- 450 m²/g
- 450 m²/g
- 450 m²/g
- 450 m²/g
- 450 m²/g
- 450 m²/g
- 450 m²/g
- 450 m²/g
- 450 m²/g
- 450 m²/g
- 450 m²/g
- 450 m²/g
- 450 m²/g
- 450 m²/g
- 450 m²/g
- 450 m²/g
- 450 m²/g
- USP code:
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1