Standard Roller Apparatus, Wheaton®, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
Roller apparatus is used for the large scale cell culture.
- Smooth ramping digitally controlled rotation accurate to 0.01 RPM
- Maintenance free precision brushless motor
- Bright digital display for easy operation
- Multiple decking and capacity configurations available for process flexibility
Spécifications
- Catalog No:
- 89369-038
- 89369-036
- 89369-042
- 89369-040
- 89369-030
- 89369-034
- 89369-032
- 89369-024
- 89369-068
- 89369-022
- 89369-066
- 89369-028
- 89369-026
- 89369-016
- 89369-060
- 89369-014
- 89369-058
- 89369-020
- 89369-018
- 89369-062
- 89369-052
- 89369-050
- 89369-056
- 89369-054
- 89369-002
- 89369-046
- 89369-000
- 89369-044
- 89369-004
- 89369-048
- Accuracy:
- 0.01 RPM
- 0.01 RPM
- 0.01 RPM
- 0.01 RPM
- 0.01 RPM
- 0.01 RPM
- 0.01 RPM
- 0.01 RPM
- 0.01 RPM
- 0.01 RPM
- 0.01 RPM
- 0.01 RPM
- 0.01 RPM
- 0.01 RPM
- 0.01 RPM
- 0.01 RPM
- 0.01 RPM
- 0.01 RPM
- 0.01 RPM
- 0.01 RPM
- 0.01 RPM
- 0.01 RPM
- 0.01 RPM
- 0.01 RPM
- 0.01 RPM
- 0.01 RPM
- 0.01 RPM
- 0.01 RPM
- 0.01 RPM
- 0.01 RPM
- Drive:
- Belt Driven
- Belt Driven
- Belt Driven
- Belt Driven
- Belt Driven
- Belt Driven
- Belt Driven
- Belt Driven
- Belt Driven
- Belt Driven
- Belt Driven
- Belt Driven
- Belt Driven
- Belt Driven
- Belt Driven
- Belt Driven
- Belt Driven
- Belt Driven
- Belt Driven
- Belt Driven
- Belt Driven
- Belt Driven
- Belt Driven
- Belt Driven
- Belt Driven
- Belt Driven
- Belt Driven
- Belt Driven
- Belt Driven
- Belt Driven
- Motor:
- Brushless DC
- Brushless DC
- Brushless DC
- Brushless DC
- Brushless DC
- Brushless DC
- Brushless DC
- Brushless DC
- Brushless DC
- Brushless DC
- Brushless DC
- Brushless DC
- Brushless DC
- Brushless DC
- Brushless DC
- Brushless DC
- Brushless DC
- Brushless DC
- Brushless DC
- Brushless DC
- Brushless DC
- Brushless DC
- Brushless DC
- Brushless DC
- Brushless DC
- Brushless DC
- Brushless DC
- Brushless DC
- Brushless DC
- Brushless DC
- Electrical:
- 100–240V, 50/60 Hz, 35W
- 100–240V, 50/60 Hz, 35W
- 100–240V, 50/60 Hz, 35W
- 100–240V, 50/60 Hz, 35W
- 100–240V, 50/60 Hz, 35W
- 100–240V, 50/60 Hz, 35W
- 100–240V, 50/60 Hz, 35W
- 100–240V, 50/60 Hz, 35W
- 100–240V, 50/60 Hz, 35W
- 100–240V, 50/60 Hz, 35W
- 100–240V, 50/60 Hz, 35W
- 100–240V, 50/60 Hz, 35W
- 100–240V, 50/60 Hz, 35W
- 100–240V, 50/60 Hz, 35W
- 100–240V, 50/60 Hz, 35W
- 100–240V, 50/60 Hz, 35W
- 100–240V, 50/60 Hz, 35W
- 100–240V, 50/60 Hz, 35W
- 100–240V, 50/60 Hz, 35W
- 100–240V, 50/60 Hz, 35W
- 100–240V, 50/60 Hz, 35W
- 100–240V, 50/60 Hz, 35W
- 100–240V, 50/60 Hz, 35W
- 100–240V, 50/60 Hz, 35W
- 100–240V, 50/60 Hz, 35W
- 100–240V, 50/60 Hz, 35W
- 100–240V, 50/60 Hz, 35W
- 100–240V, 50/60 Hz, 35W
- 100–240V, 50/60 Hz, 35W
- 100–240V, 50/60 Hz, 35W
- Max. humidity:
- 80% up to 31 °C
- 80% up to 31 °C
- 80% up to 31 °C
- 80% up to 31 °C
- 80% up to 31 °C
- 80% up to 31 °C
- 80% up to 31 °C
- 80% up to 31 °C
- 80% up to 31 °C
- 80% up to 31 °C
- 80% up to 31 °C
- 80% up to 31 °C
- 80% up to 31 °C
- 80% up to 31 °C
- 80% up to 31 °C
- 80% up to 31 °C
- 80% up to 31 °C
- 80% up to 31 °C
- 80% up to 31 °C
- 80% up to 31 °C
- 80% up to 31 °C
- 80% up to 31 °C
- 80% up to 31 °C
- 80% up to 31 °C
- 80% up to 31 °C
- 80% up to 31 °C
- 80% up to 31 °C
- 80% up to 31 °C
- 80% up to 31 °C
- 80% up to 31 °C
- Max. capacity:
- 5 - 88 position
- 5 - 88 position
- 5 - 88 position
- 5 - 88 position
- 5 - 88 position
- 5 - 88 position
- 5 - 88 position
- 5 - 88 position
- 5 - 88 position
- 5 - 88 position
- 5 - 88 position
- 5 - 88 position
- 5 - 88 position
- 5 - 88 position
- 5 - 88 position
- 5 - 88 position
- 5 - 88 position
- 5 - 88 position
- 5 - 88 position
- 5 - 88 position
- 5 - 88 position
- 5 - 88 position
- 5 - 88 position
- 5 - 88 position
- 5 - 88 position
- 5 - 88 position
- 5 - 88 position
- 5 - 88 position
- 5 - 88 position
- 5 - 88 position
- W×D×H:
- 790×630×350 mm
- 790×630×1790 mm
- 790×630×650 mm
- 790×630×350 mm
- -
- 790×630×1790 mm
- -
- -
- 120×630×1870 mm
- -
- 120×630×1870 mm
- -
- -
- 790×630×340 mm
- -
- 790×630×340 mm
- -
- 790×630×740 mm
- 790×630×740 mm
- 790×630×1870 mm
- -
- -
- -
- -
- 1200×630×1900 mm
- -
- 790×630×1900 mm
- 790×630×650 mm
- 1200×630×1900 mm
- -